Learn More
Sino Biological Human S100A4 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10185H01H50
A DNA sequence encoding the human S100A4 (NP_002952.1) (Met 1-Lys 101) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4247