Learn More
Sino Biological CD45/PTPRC Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 101854MM15100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the N-terminus of the Human CD45/PTPRC. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2614