Learn More
Sino Biological CD147 / EMMPRIN / Basigin Antibody (PE), Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10186R125P100
This antibody was obtained from a rabbit immunized with purified, recombinant Human WFDC2 / HE4 (rh WFDC2 / HE4; Catalog#12609-H08H; NP_006094.3; Met1-Phe124) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-2651