Learn More
Sino Biological CD147 / EMMPRIN / Basigin Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10186RP01200
Produced in rabbits immunized with purified, recombinant Human CD147. (rh CD147; Catalog#10186-H08H; NP_940991.1; Met 1-His 205). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2654