Learn More
Sino Biological CD147 / EMMPRIN / Basigin Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10186RP02200
Produced in rabbits immunized with purified, recombinant Human CD147. (rhCD147; Catalog#10186-H08H; NP_940991.1; Met 1-His 205). CD147 specific IgG was purified by human CD147 affinity chromatography.
Catalog No. 50-172-2657