Learn More
Sino Biological Human ESAM / Endothelial Cell Adhesion Molecule Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10187H02H50
A DNA sequence encoding the extracellular domain (Met 1-Ala 248) of human ESAM (NP_620411.2) precursor was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4257