Learn More
Sino Biological Human ESAM / Endothelial Cell Adhesion Molecule Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10187H08H50
A DNA sequence encoding the extracellular domain (Met 1-Ala 248) of human ESAM (NP_620411.2) precursor was expressed,with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4259