Learn More
Sino Biological ESAM / Endothelial Cell Adhesion Molecule Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10187RP02200
Produced in rabbits immunized with purified, recombinant Human ESAM extracellular domain (rh ESAM; Catalog#10187-H08H; NP_620411.2; Met 1-Ala 248). ESAM specific IgG was purified by human ESAM affinity chromatography.
Catalog No. 50-172-2676