Learn More
Sino Biological Human Ephrin-A3 / EFNA3 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10188H08H100
A DNA sequence encoding the human EphrinA3 (NP_004943.1) (Met 1-Ser 213) with the C-terminal propeptide removed was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4266