Learn More
Sino Biological Human Ephrin-A3 / EFNA3 Protein DFS Item
Manufacturer:  Sino Biological 10188HCCH100
A DNA sequence encoding the human EphrinA3 (NP_004943.1) (Met 1-Ser 213) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4269