Learn More
Sino Biological Ephrin-A3/EFNA3 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10188MM0850
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Ephrin-A3/EFNA3 (rh Ephrin-A3/EFNA3; Catalog#10188-H08H; NP_004943.1; Met1-Ser213). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2693