Learn More
Sino Biological CD171/L1CAM Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101901T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse CD171/L1CAM, and purified by antigen affinity chromatography.
Catalog No. 50-172-2715