Learn More
Sino Biological LY75 / DEC-205 / CD205 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101920T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse LY75 / DEC-205 / CD205, and purified by antigen affinity chromatography.
Catalog No. 50-172-2721