Learn More
Sino Biological Frizzled-4/FZD4/CD344 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101926T32100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse Frizzled-4/FZD4/CD344, and purified by antigen affinity chromatography.
Catalog No. 50-172-2732