Learn More
Sino Biological CD61 / Integrin beta 3 / ITGB3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101954T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse CD61 / Integrin beta 3 / ITGB3, and purified by antigen affinity chromatography.
Catalog No. 50-172-2751