Learn More
Sino Biological CD44 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101972T08100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse CD44, and purified by antigen affinity chromatography.
Catalog No. 50-172-2785