Learn More
Sino Biological CD131 / CSF2RB / IL3RB / IL5RB Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101978T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse CD131 / CSF2RB / IL3RB / IL5RB, and purified by antigen affinity chromatography.
Catalog No. 50-172-2789