Learn More
Sino Biological Human JAM-A / F11R Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10198H02H50
A DNA sequence encoding the extracellular domain (Met 1-Ala 242) of human JAM-A (NP_058642.1) precursor was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4293