Learn More
Sino Biological JAM-A / F11R Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10198MM0250
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human JAM-A extracellular domain (rh JAM-A; Catalog#10198-H08H; Met 1-Ala 242; NP_058642.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2823