Learn More
Sino Biological JAM-A / F11R Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10198RP01200
Produced in rabbits immunized with purified, recombinant Human JAM-A extracellular domain (rhJAM-A; Catalog#10198-H08H; Met 1-Ala 242; NP_058642.1). Total IgG was purified by Protein A affinity chromatography .
Catalog No. 50-172-2838