Learn More
Sino Biological OX-40L / TNFSF4 / CD252 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101986T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse OX-40L / TNFSF4 / CD252, and purified by antigen affinity chromatography.
Catalog No. 50-172-2818