Learn More
Sino Biological KLK13 / Kallikrein 13 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10199MM06200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human KLK13 / Kallikrein 13 (rh KLK13 / Kallikrein 13; Catalog#10199-H08H; NP_056411.1; Met1-Ile262). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2857