Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Sino Biological KLK13 / Kallikrein-13 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10199RP02200
Produced in rabbits immunized with purified, recombinant Human KLK13 / Kallikrein13 (rh KLK13; Catalog#10199-H08H; NP_056411.1; Met 1-Ile 262). KLK13 specific IgG was purified by human KLK13 affinity chromatography .
Catalog No. 50-172-2865