Learn More
Sino Biological NFKBIB / IkB-beta Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101993T42100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse NFKBIB / IkB-beta, and purified by antigen affinity chromatography.
Catalog No. 50-172-2846