Learn More
Sino Biological CD209 / DC-SIGN Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10200MM0150
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD209 / DC-SIGN. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2897