Learn More
Sino Biological CD209/DC-SIGN Antibody (PE), Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10200R037P100
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD209/DC-SIGN (rh CD209/DC-SIGN; Catalog#10200-H01H; NP_066978.1; Lys62-Ala404) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-2910