Learn More
Sino Biological CD209 / DC-SIGN Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10200T16200
Produced in rabbits immunized with purified, recombinant Human CD209 / DC-SIGN (rh CD209 / DC-SIGN; Catalog#10200-H01H; NP_066978.1; Lys62-Ala404). CD209 / DC-SIGN specific IgG was purified by Human CD209 / DC-SIGN affinity chromatography.
Catalog No. 50-172-2921