Learn More
Sino Biological CD45/PTPRC Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102001T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CD45/PTPRC, and purified by antigen affinity chromatography.
Catalog No. 50-172-2871