Learn More
Sino Biological DPP4 / CD26 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102004T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DPP4 / CD26, and purified by antigen affinity chromatography.
Catalog No. 50-172-2880