Learn More
Sino Biological Insulin Receptor/INSR/CD220 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102005T1050
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Insulin Receptor/INSR/CD220, and purified by antigen affinity chromatography.
Catalog No. 50-172-2884