Learn More
Sino Biological OX40/TNFRSF4/CD134 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102007T36200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Rat OX40/TNFRSF4/CD134, and purified by antigen affinity chromatography.
Catalog No. 50-172-2892