Learn More
Sino Biological Human HER3 / ErbB3 Protein DFS Item
Manufacturer:  Sino Biological 10201HCCH50
A DNA sequence encoding the human ERBB3 (NP_001973.2) (Met1-Thr643) was expressed with five amino acids (DDDDK) at the C-terminus.
Catalog No. 50-161-4311