Learn More
Sino Biological Fas Ligand / FASLG / CD95L Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102013T34200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Rat Fas Ligand / FASLG / CD95L, and purified by antigen affinity chromatography.
Catalog No. 50-172-2927