Learn More
Sino Biological Leptin Receptor / CD295 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102018T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Rat Leptin Receptor / CD295, and purified by antigen affinity chromatography.
Catalog No. 50-172-2937