Learn More
Sino Biological Dkk3 / REIC Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10203RP0450
Produced in rabbits immunized with purified, recombinant Human Dkk3 (rhDkk3; Catalog#10203-H08H; aa 1-350; NP_001018067.1). Dkk3 specific IgG was purified by human Dkk3 affinity chromatography.
Catalog No. 50-172-3007