Learn More
Sino Biological Human E-Cadherin / CDH1 / E-cad / CD324 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10204H02H50
A DNA sequence encoding the human E-Cad (P12830)(Met1-Ile707) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4321