Learn More
Sino Biological E-Cadherin / CDH1 / E-cad / CD324 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10204RP01200
Produced in rabbits immunized with purified, recombinant Human Cadherin-1 / CDH1 (rh Cadherin-1; Catalog#10204-H08H; NP_004351.1; Met 1-Ile 707). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-3045