Learn More
Sino Biological E-Cadherin/CDH1/E-cad/CD324 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10204T26200
Produced in rabbits immunized with purified, recombinant Human E-Cadherin/CDH1/E-cad/CD324 (rh E-Cadherin/CDH1/E-cad/CD324; Catalog#10204-H08H; NP_004351.1; Met1-Ile707). E-Cadherin/CDH1/E-cad/CD324 specific IgG was purified by Human E-Cadherin/CDH1/E-cad/CD324 affinity chromatography.
Catalog No. 50-172-3048