Learn More
Sino Biological CYP2E1 / Cytochrome P450 2E1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102050T4050
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CYP2E1 / Cytochrome P450 2E1, and purified by antigen affinity chromatography.
Catalog No. 50-172-3055