Learn More
Sino Biological Fibrillarin/FBL Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102070T38200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Fibrillarin/FBL, and purified by antigen affinity chromatography.
Catalog No. 50-172-3087