Learn More
Sino Biological Human Layilin / LAYN Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10208H02H20
A DNA sequence encoding the extracellular domain (Met 1-Glu 220) of human Layilin (NP_849156.1) precursor was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4335