Learn More
Sino Biological Layilin / LAYN Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10208MM08200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Layilin / LAYN (rh Layilin / LAYN; Catalog#10208-H08H; NP_849156.1; Met1-Glu220). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3116