Learn More
Sino Biological PARK2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102096T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PARK2, and purified by antigen affinity chromatography.
Catalog No. 50-172-3137