Learn More
Sino Biological LMNA/lamins A Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102105T3450
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human LMNA/lamins A, and purified by antigen affinity chromatography.
Catalog No. 50-172-3149