Learn More
Sino Biological Human Latexin / LXN / TCI Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10211H07E20
A DNA sequence encoding the human Latexin (NP_064554.3) (Glu 2-Glu222) was expressed, fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4349