Learn More
Sino Biological Latexin / Lxn Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10211MM02200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, E.coli-derived, recombinant human Latexin (rh Latexin; Catalog#10211-H07E; NP_064554.3; Glu 2-Glu 222). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3160