Learn More
Sino Biological Latexin / LXN / TCI Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10211RP0250
Produced in rabbits immunized with purified, recombinant Human Latexin (rh Latexin; Catalog#10211-H07E; NP_064554.3; Glu 2-Glu 222). Latexin specific IgG was purified by human Latexin affinity chromatography.
Catalog No. 50-172-3180