Learn More
Sino Biological ZAP Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102124T3650
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ZAP, and purified by antigen affinity chromatography.
Catalog No. 50-172-3192