Learn More
Sino Biological Human BMP-5 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10214H01H10
A DNA sequence encoding the carboxy-terminal domain (Gln 324-His 454) (the mature chain) of human BMP5 (NP_066551.1) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4354