Learn More
Sino Biological Nlrp3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102161T0250
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nlrp3, and purified by antigen affinity chromatography.
Catalog No. 50-172-3245